Assay Details
Target Gene Details
Entrez Gene ID: | 11634 |
Gene Name: | autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy) |
Gene Aliases: |
- |
Location: |
Chr.10:78030022-78043610 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aire | NM_001271549.1 | 1 | 88 | NP_001258478.1 |
NM_001271550.1 | 1 | 88 | NP_001258479.1 | |
NM_001271551.1 | 1 | 88 | NP_001258480.1 | |
NM_001271552.1 | 1 | 88 | NP_001258481.1 | |
NM_001271553.1 | 1 | 88 | NP_001258482.1 | |
NM_001271554.1 | 1 | 88 | NP_001258483.1 | |
NM_001271555.1 | 1 | 88 | NP_001258484.1 | |
NM_001271556.1 | 1 | 88 | NP_001258485.1 | |
NM_001271557.1 | 1 | 88 | NP_001258486.1 | |
NM_001271558.1 | 1 | 88 | NP_001258487.1 | |
NM_001271559.1 | 1 | 88 | NP_001258488.1 | |
NM_009646.2 | 1 | 88 | NP_033776.1 | |
NR_073358.1 | 1 | 88 | ||
AF079536.1 | AAD20444.1 | |||
AF128115.1 | AAF36460.1 | |||
AF128116.1 | AAF36461.1 | |||
AF128117.1 | AAF36462.1 | |||
AF128118.1 | AAF36463.1 | |||
AF128119.1 | AAF36464.1 | |||
AF128120.1 | AAF36465.1 | |||
AF128121.1 | AAF36466.1 | |||
AF128122.1 | AAF36467.1 | |||
AF128123.1 | AAF36468.1 | |||
AF128124.1 | AAF36469.1 | |||
AF128125.1 | AAF36470.1 | |||
AF128772.1 | AAF36481.1 | |||
AF128773.1 | AAF36482.1 | |||
AJ132243.1 | CAB36909.1 | |||
AJ243821.1 | CAB66141.1 | |||
AK153690.1 | 1 | 89 | ||
BC103510.1 | AAI03511.1 | |||
BC103511.1 | AAI03512.1 | |||
BC103518.1 | AAI03519.1 | |||
EU625343.3 | ||||
JF267399.1 |
Target Gene Details
Entrez Gene ID: | 54427 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 3-like |
Gene Aliases: |
D6Ertd14e, ecat7 |
Location: |
Chr.10:78042287-78063622 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnmt3l | NM_001081695.2 | NP_001075164.1 | ||
EF051624.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |