Assay Details
Target Gene Details
Entrez Gene ID: | 74048 |
Gene Name: | RIKEN cDNA 4632428N05 gene |
Gene Aliases: |
Dies1, PD-1H, VISTA |
Location: |
Chr.10:60346851-60428381 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4632428N05Rik | NM_001159572.1 | NP_001153044.1 | ||
NM_028732.4 | NP_083008.1 | |||
AK014600.1 | BAB29455.1 | |||
AK030479.1 | BAC26981.1 | |||
AK032383.1 | BAC27847.1 | |||
AK033393.1 | ||||
AK079734.1 | BAC37735.1 | |||
AK131118.1 | BAD21368.1 | |||
AK159086.1 | BAE34804.1 | |||
AK159864.1 | BAE35438.1 | |||
AK163118.1 | BAE37201.1 | |||
AK170059.1 | BAE41539.1 | |||
AK170419.1 | BAE41785.1 | |||
AK171045.1 | ||||
BC003967.1 | AAH03967.1 | |||
EF426799.1 | ||||
JN602184.1 |
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Intron 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh23 | NM_001252635.1 | NP_001239564.1 | ||
NM_023370.3 | NP_075859.2 | |||
NR_045556.1 | ||||
AF308939.1 | AAG52817.1 | |||
AJ888000.1 | CAI59609.1 | |||
AJ888001.1 | CAI59610.1 | |||
AJ888002.1 | CAI59611.1 | |||
AY026062.1 | AAK07670.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |