Assay Details
Target Gene Details
Entrez Gene ID: | 215814 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
1700009P13Rik, AI480677 |
Location: |
Chr.10:18213685-18234991 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc28a | NM_144820.3 | 4 | 612 | NP_659069.2 |
AK028075.1 | 4 | 581 | BAC25738.1 | |
AK046998.1 | 2 | 236 | BAC32937.1 | |
AK148447.1 | 4 | 613 | BAE28559.1 | |
BC027759.1 | 4 | 620 |
More Information
Set Membership: |
Intragenic Exonic |