Assay Details
Target Gene Details
Entrez Gene ID: | 215821 |
Gene Name: | ARFGEF family member 3 |
Gene Aliases: |
9630004D11, B930094H20Rik, BIG3, D10Bwg1379e, mKIAA1244 |
Location: |
Chr.10:18581839-18744069 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arfgef3 | NM_001033258.4 | NP_001028430.1 | ||
AK122475.1 | BAC65757.1 | |||
AK147679.1 | BAE28069.1 | |||
AK147963.1 | ||||
AK148343.1 | ||||
AK165602.1 |
More Information
Set Membership: |
Intragenic Non-exonic |