Assay Details
Target Gene Details
Entrez Gene ID: | 235028 |
Gene Name: | zinc finger protein 426 |
Gene Aliases: |
2900057C04Rik, KRAB1, Zfo61, Zfp68-rs1, Znf426, mKIAA4237 |
Location: |
Chr.9:20468548-20493922 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp426 |