Assay Details
Target Gene Details
Entrez Gene ID: | 22688 |
Gene Name: | zinc finger protein 26 |
Gene Aliases: |
5033428C05Rik, KRAB15, Zfp-26, Zfp70, Zfp81-rs1, mKIAA4196, mkr-3, mszf14, mszf52 |
Location: |
Chr.9:20428318-20460169 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp26 | NM_011753.3 | NP_035883.2 | ||
AK029298.1 | BAC26381.1 | |||
AK082002.1 | BAC38392.1 | |||
AK085739.1 | 4 | 3143 | ||
AK141740.1 | BAE24816.1 |
More Information
Set Membership: |
Intragenic Non-exonic |