Assay Details
Target Gene Details
Entrez Gene ID: | 22688 |
Gene Name: | zinc finger protein 26 |
Gene Aliases: |
5033428C05Rik, KRAB15, Zfp-26, Zfp70, Zfp81-rs1, mKIAA4196, mkr-3, mszf14, mszf52 |
Location: |
Chr.9:20428318-20460169 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp26 | NM_011753.3 | 2 | 281 | NP_035883.2 |
AK017201.1 | 2 | 111 | ||
AK029298.1 | 2 | 105 | BAC26381.1 | |
AK082002.1 | 2 | 114 | BAC38392.1 | |
AK085739.1 | 2 | 209 | ||
AK141740.1 | 2 | 118 | BAE24816.1 | |
CA871137.1 | 2 | 281 |
More Information
Set Membership: |
Intragenic Exonic |