Assay Details
Target Gene Details
Entrez Gene ID: | 57276 |
Gene Name: | V-set and immunoglobulin domain containing 2 |
Gene Aliases: |
1190004B15Rik, 2210413P10Rik, AU040484, CTX, Ctm |
Location: |
Chr.9:37538814-37545123 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vsig2 | NM_020518.2 | 3 | 445 | NP_065264.2 |
AK004478.1 | BAB23323.1 | |||
AK008920.1 | 3 | 407 | BAB25968.1 | |
BC099457.1 | 3 | 409 | AAH99457.1 |
More Information
Set Membership: |
Intragenic Exonic |