Assay Details
Target Gene Details
Entrez Gene ID: | 22040 |
Gene Name: | three prime repair exonuclease 1 |
Gene Aliases: |
1661, AU041952 |
Location: |
Chr.9:109057932-109059723 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trex1 | NM_001012236.1 | NP_001012236.1 | ||
NM_011637.6 | NP_035767.4 | |||
AF151106.3 | AAD48775.2 | |||
AF319574.1 | 1 | 533 | AAK07621.1 | |
AK009899.1 | BAB26571.2 | |||
AK087832.1 | BAC40020.1 | |||
AK171916.1 | BAE42731.1 | |||
BC011133.1 | AAH11133.1 | |||
BY234700.1 | ||||
CA323364.1 |