Assay Details
Target Gene Details
Entrez Gene ID: | 21667 |
Gene Name: | teratocarcinoma-derived growth factor 1 |
Gene Aliases: |
CR1, cripto |
Location: |
Chr.9:110939608-110946158 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tdgf1 | NM_011562.2 | NP_035692.2 | ||
AK133730.1 | BAE21807.1 | |||
AK163969.1 | ||||
BC052646.1 | AAH52646.1 | |||
M87321.1 | AAA37459.1 |
More Information
Set Membership: |
Intragenic Non-exonic |