Assay Details
Target Gene Details
Entrez Gene ID: | 69519 |
Gene Name: | RWD domain containing 2A |
Gene Aliases: |
1700030C20Rik, AI848608, Rwdd2 |
Location: |
Chr.9:86571988-86574899 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rwdd2a | NM_001145968.1 | NP_001139440.1 | ||
NM_001145969.1 | NP_001139441.1 | |||
NM_027100.2 | NP_081376.2 | |||
AK006533.1 | BAB24638.1 | |||
BC115691.1 | ||||
BC115692.1 | ||||
CJ149958.1 | ||||
CN836837.1 |
More Information
Set Membership: |
Intragenic Exonic |