Assay Details
Target Gene Details
Entrez Gene ID: | 234967 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 4 |
Gene Aliases: |
6330573I15Rik, PAT4 |
Location: |
Chr.9:15709769-15742388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc36a4 | NM_172289.4 | NP_758493.2 | ||
AF453746.1 | AAN76274.1 | |||
AK032074.1 | BAC27683.1 | |||
AK052277.1 | ||||
AK150167.1 | ||||
BC115964.1 |
More Information
Set Membership: |
Intragenic Non-exonic |