Assay Details
Target Gene Details
Entrez Gene ID: | 408059 |
Gene Name: | cDNA sequence BC049352 |
Gene Aliases: |
- |
Location: |
Chr.9:45181508-45251370 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BC049352 | AK138393.1 |
Target Gene Details
Entrez Gene ID: | 214523 |
Gene Name: | transmembrane protease, serine 4 |
Gene Aliases: |
mCAP2 |
Location: |
Chr.9:45172726-45204075 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmprss4 | NM_145403.2 | NP_663378.1 | ||
AK078890.1 | ||||
AY043240.1 | AAK85307.1 | |||
BC021368.1 | AAH21368.1 |
More Information
Set Membership: |
Intragenic Non-exonic |