Assay Details
Target Gene Details
Entrez Gene ID: | 19035 |
Gene Name: | peptidylprolyl isomerase B |
Gene Aliases: |
AA408962, AA553318, AI844835, Cphn-2, Cphn2, CyP-20b |
Location: |
Chr.9:66060169-66066629 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppib | NM_011149.2 | 1 | 154 | NP_035279.2 |
AK002357.1 | 1 | 101 | BAB22036.1 | |
AK128977.1 | 1 | 104 | BAC87663.1 | |
AK154073.1 | 1 | 93 | BAE32357.1 | |
AL363449.1 | 1 | 195 | ||
BC013061.1 | 1 | 91 | AAH13061.1 | |
M60456.1 | 1 | 73 | AAA37498.1 | |
X58990.1 | 1 | 199 | CAA41736.1 |
More Information
Set Membership: |
Intragenic Exonic |