Assay Details
Target Gene Details
Entrez Gene ID: | 59092 |
Gene Name: | poly(rC) binding protein 4 |
Gene Aliases: |
1200003L19Rik, AlphaCP-4 |
Location: |
Chr.9:106453406-106464012 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcbp4 | NM_021567.5 | NP_067542.3 | ||
AF176328.1 | AAG09239.1 | |||
AK004580.1 | ||||
AK005383.1 | BAB23990.1 | |||
AK077632.1 | BAC36915.1 | |||
AK082005.1 | ||||
BC010694.1 | AAH10694.1 | |||
BY259572.1 | ||||
CJ162439.1 | ||||
CJ174803.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |