Assay Details
Target Gene Details
Entrez Gene ID: | 258601 |
Gene Name: | olfactory receptor 984 |
Gene Aliases: |
MOR239-6 |
Location: |
Chr.9:40100544-40156029 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Olfr984 |
Target Gene Details
Entrez Gene ID: | 75667 |
Gene Name: | transmembrane protein 225 |
Gene Aliases: |
1700030E15Rik, Tnen225 |
Location: |
Chr.9:40148100-40150901 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem225 | NM_029379.1 | NP_083655.1 | ||
AK006538.1 | BAB24641.1 | |||
BC116869.1 | ||||
BC116871.1 |