Assay Details
Target Gene Details
Entrez Gene ID: | 22273 |
Gene Name: | ubiquinol-cytochrome c reductase core protein 1 |
Gene Aliases: |
1110032G10Rik |
Location: |
Chr.9:108936648-108949641 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uqcrc1 | NM_025407.2 | NP_079683.2 | ||
AI462428.1 | ||||
AK002292.1 | ||||
AK004031.1 | ||||
AK010553.1 | BAB27022.1 | |||
AK013128.1 | BAB28666.1 | |||
AK151764.1 | BAE30670.1 | |||
AK160337.1 | BAE35744.1 | |||
AK167911.1 | BAE39918.1 | |||
AK168218.1 | BAE40175.1 | |||
AK189225.1 | ||||
BC018405.1 | AAH18405.1 | |||
BC030064.1 | AAH30064.1 |
More Information
Set Membership: |
Intragenic Non-exonic |