Assay Details
Target Gene Details
Entrez Gene ID: | 382083 |
Gene Name: | sorting nexin 22 |
Gene Aliases: |
Gm1120 |
Location: |
Chr.9:66065176-66069780 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snx22 | NM_001025612.2 | NP_001020783.1 | ||
AK032028.1 | BAC27659.1 | |||
BC138950.1 | ||||
BC138951.1 |
More Information
Set Membership: |
Intragenic Non-exonic |