Assay Details
Target Gene Details
Entrez Gene ID: | 235459 |
Gene Name: | general transcription factor II A, 2 |
Gene Aliases: |
TFIIA-gamma, TfIIg, Tfiia2 |
Location: |
Chr.9:70011851-70022866 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2a2 | NM_001039519.2 | NP_001034608.1 | ||
NM_001282083.1 | NP_001269012.1 | |||
NM_001282084.1 | NP_001269013.1 | |||
NR_104093.1 | ||||
AK050621.1 | BAE20680.1 | |||
AK139783.1 | BAE24136.1 | |||
AK164255.1 | ||||
AK165466.1 | BAE38203.1 | |||
AK169778.1 | BAE41360.1 | |||
AK212444.1 | ||||
BB842876.1 | ||||
BC028551.1 | ||||
BG088284.2 |
More Information
Set Membership: |
Intragenic Non-exonic |