Assay Details
Target Gene Details
Entrez Gene ID: | 64011 |
Gene Name: | neurogranin |
Gene Aliases: |
0710001B06Rik, AI838505, NG, NG/RC3, Pss1, R75334, RC3 |
Location: |
Chr.9:37544493-37552745 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrgn | NM_022029.2 | 2 | 247 | NP_071312.1 |
AK002933.1 | 2 | 207 | BAB22466.1 | |
AK158630.1 | 2 | 234 | BAE34589.1 | |
BC061102.1 | 2 | 211 | AAH61102.1 | |
BC138511.1 | 2 | 124 | ||
BC138513.1 | 2 | 124 | ||
BU604757.1 | 2 | 250 |