Assay Details
Target Gene Details
Entrez Gene ID: | 23873 |
Gene Name: | Fas apoptotic inhibitory molecule |
Gene Aliases: |
- |
Location: |
Chr.9:98986373-99002019 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Faim | NM_001122851.1 | NP_001116323.1 | ||
NM_011810.3 | NP_035940.3 | |||
AK005762.1 | BAB24225.1 | |||
AK156341.1 | BAE33681.1 | |||
BY055691.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)