Assay Details
Target Gene Details
Entrez Gene ID: | 235606 |
Gene Name: | acylpeptide hydrolase |
Gene Aliases: |
- |
Location: |
Chr.9:108085413-108094548 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apeh | NM_146226.2 | NP_666338.2 | ||
AK149671.1 | ||||
AK149713.1 | ||||
AK178617.1 | ||||
AK182596.1 | ||||
BB737329.1 | ||||
BC025494.1 | AAH25494.1 | |||
BC026594.1 | AAH26594.1 | |||
BC034199.1 | AAH34199.1 |
More Information
Set Membership: |
Intragenic Non-exonic |