Assay Details
Target Gene Details
Entrez Gene ID: | 12539 |
Gene Name: | cell division cycle 37 |
Gene Aliases: |
p50, p50Cdc37 |
Location: |
Chr.9:21138541-21149977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdc37 | NM_016742.4 | NP_058022.1 | ||
AK010494.1 | BAB26984.1 | |||
AK013255.1 | BAB28749.1 | |||
AK145671.1 | BAE26580.1 | |||
AK168651.1 | BAE40508.1 | |||
BC060079.1 | AAH60079.1 | |||
U43076.1 | AAB18761.1 |
More Information
Set Membership: |
Intragenic Non-exonic |