Assay Details
Target Gene Details
Entrez Gene ID: | 100529079 |
Gene Name: | RIKEN cDNA 5830418P13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:103423578-103461260 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
5830418P13Rik | NR_040466.1 | |||
AK079871.1 | ||||
AK134047.1 |
Target Gene Details
Entrez Gene ID: | 107993 |
Gene Name: | beaded filament structural protein 2, phakinin |
Gene Aliases: |
AI448593, CP49 |
Location: |
Chr.9:103424924-103480414 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bfsp2 | NM_001002896.2 | NP_001002896.1 | ||
AK134810.1 | BAE22294.1 | |||
AK193757.1 | ||||
BC062815.1 | AAH62815.1 | |||
BC068172.1 | AAH68172.1 | |||
S55549.1 | AAB25419.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |