Assay Details
Target Gene Details
Entrez Gene ID: | 29873 |
Gene Name: | chondroitin sulfate proteoglycan 5 |
Gene Aliases: |
Caleb, Ngc |
Location: |
Chr.9:110243783-110262576 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cspg5 | NM_001166273.1 | NP_001159745.1 | ||
NM_013884.3 | NP_038912.3 | |||
AF133700.1 | AAF23362.1 | |||
AF461090.1 | AAQ04777.1 | |||
AF461091.1 | AAQ04778.1 | |||
AF461092.1 | AAQ04779.1 | |||
AK053891.1 | BAC35578.1 | |||
BC055736.1 | AAH55736.1 |