Assay Details
Target Gene Details
Entrez Gene ID: | 67213 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 6 |
Gene Aliases: |
2810051A14Rik, AA536733, Cklfsf6 |
Location: |
Chr.9:114731157-114749347 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm6 | NM_026036.3 | NP_080312.1 | ||
AK012941.1 | BAB28562.1 | |||
AK030885.1 | BAC27172.1 | |||
AK128960.1 | ||||
AK145743.1 | BAE26621.1 | |||
AK150775.1 | ||||
AK151092.1 | ||||
AK151431.1 | ||||
AK152448.1 | BAE31227.1 | |||
AK152723.1 | BAE31446.1 | |||
AK152817.1 | BAE31520.1 | |||
AK153092.1 | ||||
AK154481.1 | BAE32617.1 | |||
AK159439.1 | BAE35085.1 | |||
AK159735.1 | BAE35329.1 | |||
AK165948.1 | BAE38480.1 | |||
AK167499.1 | BAE39575.1 | |||
AK167854.1 | BAE39872.1 | |||
AK170905.1 | BAE42106.1 | |||
AK171394.1 | BAE42431.1 | |||
AY241868.1 | AAP33490.1 | |||
BC027248.1 | AAH27248.1 |
More Information
Set Membership: |
Intragenic Non-exonic |