Assay Details
Target Gene Details
Entrez Gene ID: | 12769 |
Gene Name: | chemokine (C-C motif) receptor 9 |
Gene Aliases: |
A130091K22Rik, Cmkbr10, GPR-9-6 |
Location: |
Chr.9:123767211-123783457 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr9 | NM_001166625.1 | NP_001160097.1 | ||
NM_009913.6 | NP_034043.1 | |||
AJ131357.2 | CAB66136.2 | |||
AJ132336.1 | CAB43480.1 | |||
AK050615.1 | BAC34344.1 | |||
AK153975.1 | BAE32291.1 | |||
BC132166.1 | ||||
BC132466.1 |