Assay Details
Target Gene Details
Entrez Gene ID: | 66167 |
Gene Name: | translational machinery associated 7 homolog (S. cerevisiae) |
Gene Aliases: |
1110017O22Rik, Ccdc72 |
Location: |
Chr.9:109077988-109082381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tma7 | NM_183250.2 | NP_899073.1 | ||
AK002863.1 | ||||
AK003762.1 | BAC25052.1 | |||
AK160705.1 | BAE35964.1 | |||
AK169860.1 | BAE41416.1 | |||
BC106104.1 | AAI06105.1 | |||
BC116987.1 | ||||
BC116989.1 |