Assay Details
Target Gene Details
Entrez Gene ID: | 66113 |
Gene Name: | apolipoprotein A-V |
Gene Aliases: |
1300007O05Rik, Apoav, RAP3 |
Location: |
Chr.9:46268608-46271919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoa5 | NM_080434.3 | NP_536682.2 | ||
AF327059.1 | AAG49600.1 | |||
AK004903.1 | ||||
AK004936.1 | ||||
AK050280.1 | BAC34163.1 | |||
BC011198.1 | AAH11198.1 |
More Information
Set Membership: |
Intragenic Non-exonic |