Assay Details
Target Gene Details
Entrez Gene ID: | 67789 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
6330580J24Rik, C77829, mir-425 |
Location: |
Chr.9:108567496-108572773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dalrd3 | NM_026378.3 | NP_080654.3 | ||
AK002439.1 | BAC24990.1 | |||
AK149963.1 | BAE29197.1 | |||
AK169405.1 | BAE41151.1 | |||
AK171818.1 | BAE42679.1 | |||
AK195213.1 | ||||
AK206691.1 | ||||
BC013483.1 | AAH13483.2 |