Assay Details
Target Gene Details
Entrez Gene ID: | 94187 |
Gene Name: | zinc finger protein 423 |
Gene Aliases: |
Ebfaz, Roaz, Zfp104, Znf423, ataxia1, mKIAA0760, nur12 |
Location: |
Chr.8:87661810-87961859 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp423 | NM_033327.2 | NP_201584.2 | ||
AK122365.1 | BAC65647.1 | |||
AY256893.1 | AAP33073.1 | |||
BC139028.1 | ||||
BC139030.1 | ||||
BQ180174.1 | ||||
CL210542.2 |
More Information
Set Membership: |
Intragenic Non-exonic |