Assay Details
Target Gene Details
Entrez Gene ID: | 20516 |
Gene Name: | solute carrier family 20, member 2 |
Gene Aliases: |
MolPit2, Pit-2, Pit2, Ram-1, Ram1 |
Location: |
Chr.8:22476700-22569616 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc20a2 | NM_011394.3 | 7 | 1284 | NP_035524.2 |
AF196476.1 | 6 | 1052 | AAG28493.1 | |
AK087507.1 | 7 | 1109 | ||
AK157206.1 | 7 | 1158 | BAE33999.1 | |
AK170985.1 | 7 | 1250 | BAE42159.1 | |
AK192413.1 | ||||
AK198610.1 | ||||
AK216704.1 | 3 | 295 | ||
AK220047.1 | ||||
BC046510.1 | 7 | 1207 | AAH46510.1 | |
CF949043.1 | 1 | 75 |
More Information
Set Membership: |
Intragenic Exonic |