Assay Details
Target Gene Details
Entrez Gene ID: | 20516 |
Gene Name: | solute carrier family 20, member 2 |
Gene Aliases: |
MolPit2, Pit-2, Pit2, Ram-1, Ram1 |
Location: |
Chr.8:22476700-22569616 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc20a2 | NM_011394.3 | 6 | 1166 | NP_035524.2 |
AF196476.1 | 5 | 934 | AAG28493.1 | |
AK087507.1 | 6 | 991 | ||
AK157206.1 | 6 | 1040 | BAE33999.1 | |
AK170985.1 | 6 | 1132 | BAE42159.1 | |
AK192413.1 | 3 | 205 | ||
AK198610.1 | 4 | 330 | ||
AK216704.1 | 2 | 177 | ||
AK220047.1 | 3 | 195 | ||
BC046510.1 | 6 | 1089 | AAH46510.1 | |
U62559.1 | 5 | 645 | AAB06046.1 |
More Information
Additional Information:
Set Membership: |
Intragenic Exonic Validated |