Assay Details
Target Gene Details
Entrez Gene ID: | 20516 |
Gene Name: | solute carrier family 20, member 2 |
Gene Aliases: |
MolPit2, Pit-2, Pit2, Ram-1, Ram1 |
Location: |
Chr.8:22476700-22569616 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc20a2 | NM_011394.3 | 3 | 849 | NP_035524.2 |
AF196476.1 | 2 | 617 | AAG28493.1 | |
AK076380.1 | 1 | 94 | ||
AK087507.1 | 3 | 673 | ||
AK157206.1 | 3 | 723 | BAE33999.1 | |
AK170985.1 | 3 | 815 | BAE42159.1 | |
AK177809.1 | 2 | 131 | ||
AK181075.1 | 2 | 131 | ||
AK198422.1 | 2 | 131 | ||
AK198610.1 | ||||
AK205342.1 | 2 | 131 | ||
AK208287.1 | 2 | 131 | ||
AK211983.1 | 2 | 131 | ||
AK217052.1 | 2 | 131 | ||
AK217778.1 | 2 | 131 | ||
BC046510.1 | 3 | 772 | AAH46510.1 | |
U62559.1 | 2 | 328 | AAB06046.1 |
More Information
Additional Information:
Set Membership: |
Intragenic Exonic Validated |