Assay Details
Target Gene Details
Entrez Gene ID: | 18709 |
Gene Name: | phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta) |
Gene Aliases: |
p85beta |
Location: |
Chr.8:70768178-70776728 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pik3r2 | NM_008841.2 | NP_032867.2 | ||
AK146195.1 | BAE26969.1 | |||
AK148122.1 | ||||
BC006796.1 | AAH06796.1 | |||
BC085501.1 | AAH85501.1 | |||
Y13569.1 | CAA73903.1 |
More Information
Set Membership: |
Intragenic Non-exonic |