Assay Details
Target Gene Details
Entrez Gene ID: | 210801 |
Gene Name: | unc-5 homolog D (C. elegans) |
Gene Aliases: |
D930029E11Rik, Unc5h4, mKIAA1777 |
Location: |
Chr.8:28646717-29219682 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc5d | NM_153135.3 | NP_694775.1 | ||
AJ487854.1 | CAD32252.1 | |||
BC137615.1 | ||||
BC144783.1 | ||||
BC144784.1 | ||||
BC145695.1 | ||||
FJ824810.1 |
More Information
Set Membership: |
Intragenic Non-exonic |