Assay Details
Target Gene Details
Entrez Gene ID: | 210801 |
Gene Name: | unc-5 homolog D (C. elegans) |
Gene Aliases: |
D930029E11Rik, Unc5h4, mKIAA1777 |
Location: |
Chr.8:28646717-29219682 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc5d | NM_153135.3 | 13 | 2161 | NP_694775.1 |
AJ487854.1 | 13 | 2162 | CAD32252.1 | |
AK220417.1 | 3 | 502 | BAD90463.1 | |
BC137615.1 | 13 | 1952 | ||
BC144783.1 | 11 | 1769 | ||
BC144784.1 | 11 | 1730 | ||
BC145695.1 | 10 | 1736 | ||
FJ824810.1 | 12 | 2040 |
More Information
Set Membership: |
Intragenic Exonic |