Assay Details
Target Gene Details
Entrez Gene ID: | 69724 |
Gene Name: | ribonuclease H2, large subunit |
Gene Aliases: |
2400006P09Rik, RNASEHI, RNHIA, RNHL |
Location: |
Chr.8:84956610-84969375 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnaseh2a | NM_027187.3 | NP_081463.1 | ||
AK010292.1 | BAB26828.1 | |||
AK146518.1 | BAE27230.1 | |||
BC029252.1 | ||||
BC038158.1 | AAH38158.1 |
More Information
Set Membership: |
Intragenic Non-exonic |