Assay Details
Target Gene Details
Entrez Gene ID: | 72341 |
Gene Name: | elongator acetyltransferase complex subunit 6 |
Gene Aliases: |
2610001P13Rik, 2610002I17Rik, AV159994, Tmem103 |
Location: |
Chr.9:110305192-110322102 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elp6 | NM_001081381.1 | NP_001074850.1 | ||
AK011270.1 | BAB27508.1 | |||
AK075986.1 | BAC36098.1 | |||
BC048732.1 | AAH48732.1 | |||
CJ073544.1 |