Assay Details
Target Gene Details
Entrez Gene ID: | 69479 |
Gene Name: | RIKEN cDNA 1700029J07 gene |
Gene Aliases: |
- |
Location: |
Chr.8:45939368-45975454 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700029J07Rik |
Target Gene Details
Entrez Gene ID: | 192169 |
Gene Name: | UFM1-specific peptidase 2 |
Gene Aliases: |
1810047C23Rik |
Location: |
Chr.8:45975528-45996958 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ufsp2 | NM_138668.2 | NP_619609.1 | ||
AK075795.1 | BAC35964.1 | |||
AK078309.1 | BAC37214.1 | |||
AK160164.1 | ||||
BC005503.1 | AAH05503.1 |