Assay Details
Target Gene Details
Entrez Gene ID: | 20021 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide C |
Gene Aliases: |
33kDa, RPB3, Rpo2-3, mRBP31 |
Location: |
Chr.8:94857450-94864242 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2c | NM_009090.5 | NP_033116.2 | ||
AK146763.1 | BAE27415.1 | |||
AK151270.1 | BAE30257.1 | |||
AK160053.1 | BAE35592.1 | |||
BC002023.1 | AAH02023.1 | |||
BC099548.1 | AAH99548.1 | |||
D83999.1 | BAA12205.1 |
More Information
Set Membership: |
Intragenic Non-exonic |