Assay Details
Target Gene Details
Entrez Gene ID: | 67920 |
Gene Name: | MAK16 homolog (S. cerevisiae) |
Gene Aliases: |
2600016B03Rik, AI314911, Rbm13 |
Location: |
Chr.8:31159468-31168724 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mak16 | NM_026453.3 | NP_080729.1 | ||
AK015369.1 | BAB29815.1 | |||
AK075982.1 | BAC36094.1 | |||
AK075983.1 | BAC36095.1 | |||
BC013079.1 | AAH13079.1 |
Target Gene Details
Entrez Gene ID: | 234138 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.8:31150316-31164703 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tti2 | NM_001199988.1 | NP_001186917.1 | ||
NR_103719.1 | ||||
AK045392.1 | 7 | 2872 | BAC32341.1 | |
AK084242.1 | BAC39146.1 | |||
AK139425.1 | BAE24007.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |