Assay Details
Target Gene Details
Entrez Gene ID: | 100502619 |
Gene Name: | expressed sequence AW046200 |
Gene Aliases: |
- |
Location: |
Chr.8:57651753-57663430 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AW046200 | NR_040698.1 | |||
BY742370.1 | ||||
CJ133617.1 | ||||
CX225354.1 |
Target Gene Details
Entrez Gene ID: | 108150 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 7 |
Gene Aliases: |
AI225872 |
Location: |
Chr.8:57523825-57653055 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt7 | NM_001167981.1 | 1 | 134 | NP_001161453.1 |
NM_144731.4 | 1 | 134 | NP_653332.3 | |
AF349573.1 | 1 | 58 | AAK37549.1 | |
AK030683.1 | 1 | 81 | ||
AK033427.1 | 1 | 83 | BAC28284.1 | |
AK036523.1 | 1 | 83 | BAC29461.1 | |
AK041791.1 | 1 | 111 | BAC31068.1 | |
BC052461.1 | 1 | 110 | AAH52461.1 | |
BY167413.1 | 1 | 135 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |