Assay Details
Target Gene Details
Entrez Gene ID: | 277978 |
Gene Name: | exocyst complex component 3-like |
Gene Aliases: |
C730015A04Rik, Exoc3l1 |
Location: |
Chr.8:105289923-105296962 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Exoc3l | NM_177788.4 | 1 | 99 | NP_808456.1 |
AK050100.1 | 1 | 100 | BAC34066.1 | |
BC094238.1 | 1 | 53 | AAH94238.1 |
More Information
Set Membership: |
Intragenic Exonic |