Assay Details
Target Gene Details
Entrez Gene ID: | 13909 |
Gene Name: | carboxylesterase 3B |
Gene Aliases: |
Ces31L, Ces3L, EG13909, ES31L, Es31, Gm4738 |
Location: |
Chr.8:105083730-105093929 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ces3b | NM_001159415.1 | 6 | 800 | NP_001152887.1 |
NM_144511.2 | 6 | 800 | NP_653094.2 | |
BC010812.1 | 6 | 790 | AAH10812.1 | |
BC019147.1 | 6 | 796 | AAH19147.1 |
More Information
Set Membership: |
Intragenic Exonic |