Assay Details
Target Gene Details
Entrez Gene ID: | 27967 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
5730408I11Rik, D8Wsu96e, DAN16, Scaf6 |
Location: |
Chr.8:72460483-72476675 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cherp | NM_138585.3 | NP_613051.3 | ||
AF536541.1 | AAN77182.1 | |||
AK049765.1 | ||||
AK144328.1 | ||||
BC032191.1 |
Target Gene Details
Entrez Gene ID: | 102465647 |
Gene Name: | microRNA 7068 |
Gene Aliases: |
Gm27570, mmu-mir-7068 |
Location: |
Chr.8:72470016-72470089 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mir7068 | NR_106036.1 |
More Information
Set Membership: |
Intragenic Non-exonic |