Assay Details
Target Gene Details
Entrez Gene ID: | 20300 |
Gene Name: | chemokine (C-C motif) ligand 25 |
Gene Aliases: |
A130072A22Rik, AI852536, CKb15, Scya25, TECK |
Location: |
Chr.8:4325159-4360020 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl25 | NM_009138.3 | NP_033164.1 | ||
NR_033527.1 | ||||
AJ249480.1 | CAB55762.1 | |||
AK131875.1 | BAE20845.1 | |||
AK132700.1 | BAE21309.1 | |||
AK134138.1 | BAE22030.1 | |||
BC117033.1 | ||||
BC117037.1 | ||||
DQ158256.1 | AAZ99719.1 | |||
U86357.1 | AAB69982.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |