Assay Details
Target Gene Details
Entrez Gene ID: | 69479 |
Gene Name: | RIKEN cDNA 1700029J07 gene |
Gene Aliases: |
- |
Location: |
Chr.8:45939368-45975454 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700029J07Rik | NM_001033148.3 | 2 | 1819 | NP_001028320.1 |
AK006501.1 | 3 | 413 | ||
AK156041.1 | 2 | 1821 | BAE33559.1 | |
BC107256.2 | 3 | 302 | AAI07257.1 |