Assay Details
Target Gene Details
Entrez Gene ID: | 23923 |
Gene Name: | aminoadipate aminotransferase |
Gene Aliases: |
AI875679, Aadt, KATII, Kat2, mKat-2 |
Location: |
Chr.8:60505870-60545677 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aadat | NM_011834.2 | NP_035964.1 | ||
AF072376.1 | AAD39680.1 | |||
AK075578.1 | BAC35833.1 | |||
AK144042.1 | BAE25669.1 | |||
BC012637.1 | AAH12637.1 |
More Information
Set Membership: |
Intragenic Non-exonic |