Assay Details
Target Gene Details
Entrez Gene ID: | 68865 |
Gene Name: | ARV1 homolog (yeast) |
Gene Aliases: |
1110067L22Rik, AI461928, AW121084 |
Location: |
Chr.8:124722138-124740648 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arv1 | NM_026855.4 | NP_081131.1 | ||
AK004398.1 | BAB23288.1 | |||
AK042561.1 | 3 | 464 | BAC31293.1 | |
BC119376.1 | ||||
BC120631.1 |